Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Piperazin-1-ylpropionic acid 95%

Supplier: AMBEED, INC
Danger

A486927-1G A486927-250MG A486927-5G
77292-026EA 46 USD InStock
77292-026 77292-028 77292-030
3-Piperazin-1-ylpropionic acid 95%
3-Piperazin-1-ylpropionic acid
Precaución: for Research Use Only


Formula: C₇H₁₄N₂O₂
MDL Number: MFCD03161179
CAS Number: 27245-31-4
UN: 1759
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty