Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-m-toluic acid 97%

Supplier: AMBEED, INC
Warning

A487921-100G A487921-10G A487921-1G A487921-25G A487921-5G
77279-568EA 444 USD InStock
77279-568 77279-570 77279-572 77279-574 77279-576
2-Fluoro-m-toluic acid 97%
2-Fluoro-m-toluic acid
Precaución: for Research Use Only


Formula: C₈H₇FO₂
MDL Number: MFCD00042504
CAS Number: 315-31-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty