Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-(-)-2,2'-Bis[di(3,5-di-i-propyl-4-dimethylaminophenyl)phosphino]-6,6'-dimethoxy-1,1'-biphenyl 98%

Supplier: AMBEED, INC
Warning

A490055-100MG A490055-1G A490055-250MG
77253-172EA 77 USD InStock
77253-172 77253-174 77253-176
(R)-(-)-2,2'-Bis[di(3,5-di-i-propyl-4-dimethylaminophenyl)phosphino]-6,6'-dimethoxy-1,1'-biphenyl 98%
(R)-(-)-2,2'-Bis[di(3,5-di-i-propyl-4-dimethylaminophenyl)phosphino]-6,6'-dimethoxy-1,1'-biphenyl
Precaución: for Research Use Only


Formula: C₇₀H₁₀₀N₄O₂P₂
MDL Number: MFCD09753010
CAS Number: 352655-40-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty