Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2-Bis[(2R,5R)-2,5-diisopropyl-1-phospholanyl]ethane 98%

Supplier: AMBEED, INC
Danger

A491556-100MG A491556-250MG
77253-178EA 77 USD InStock
77253-178 77253-180
1,2-Bis[(2R,5R)-2,5-diisopropyl-1-phospholanyl]ethane 98%
1,2-Bis[(2R,5R)-2,5-diisopropyl-1-phospholanyl]ethane
Precaución: for Research Use Only


Formula: C₂₂H₄₄P₂
MDL Number: MFCD08705243
CAS Number: 136705-63-0
UN: 1325
ADR: 4.1,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty