Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(E)-3-(3-Nitrophenyl)acrylic acid 97%

Supplier: AMBEED, INC
Warning

A492513-100G A492513-25G A492513-5G
77265-604EA 127 USD InStock
77265-604 77265-606 77265-608
(E)-3-(3-Nitrophenyl)acrylic acid 97%
(E)-3-(3-Nitrophenyl)acrylic acid
Precaución: for Research Use Only


Formula: C₉H₇NO₄
MDL Number: MFCD00007277
CAS Number: 1772-76-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty