Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2-Bis(hydroxymethyl)butyric acid 95%

Supplier: AMBEED, INC
Warning

A493276-100G A493276-25G A493276-500G
77253-188EA 31 USD InStock
77253-188 77253-190 77747-011
2,2-Bis(hydroxymethyl)butyric acid 95%
2,2-Bis(hydroxymethyl)butyric acid
Precaución: for Research Use Only


Formula: C₆H₁₂O₄
MDL Number: MFCD00190088
CAS Number: 10097-02-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty