Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Paeoniflorin 98%

Supplier: AMBEED, INC
Warning

A494257-100MG A494257-10MG A494257-1G A494257-1MG A494257-250MG A494257-50MG A494257-5G
77292-076EA 47 USD InStock
77292-076 77292-078 77292-080 77292-082 77292-084 77292-086 77749-010
Paeoniflorin 98%
Paeoniflorin
Precaución: for Research Use Only


Formula: C₂₃H₂₈O₁₁
MW: 480.47 g/mol
MDL Number: MFCD00869331
CAS Number: 23180-57-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty