Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2-Furoyl)piperazine 98%

Supplier: AMBEED, INC
Warning

A497413-5G A497413-25G A497413-100G
77279-668EA 23 USD InStock
77279-668 77749-020 77749-019
1-(2-Furoyl)piperazine 98%
1-(2-Furoyl)piperazine
Precaución: for Research Use Only


Formula: C₉H₁₂N₂O₂
MDL Number: MFCD00038831
CAS Number: 40172-95-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty