Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methoxycyclopropanecarboxylic acid 98%

Supplier: AMBEED, INC
Danger

A497560-100MG A497560-250MG
77265-630EA 32 USD InStock
77265-630 77265-632
1-Methoxycyclopropanecarboxylic acid 98%
1-Methoxycyclopropanecarboxylic acid
Precaución: For research use only.


Formula: C₅H₈O₃
MDL Number: MFCD19228478
CAS Number: 100683-08-7
UN: 3261
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty