Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4',4'',4'''-(Pyrene-1,3,6,8-tetrayl)tetrabenzaldehyde 97%

Supplier: AMBEED, INC
Warning

A498450-100MG A498450-250MG A498450-5G A498450-1G
77265-634EA 20 USD InStock
77265-634 77265-636 77265-638 77551-516
4,4',4'',4'''-(Pyrene-1,3,6,8-tetrayl)tetrabenzaldehyde 97%
4,4',4'',4'''-(Pyrene-1,3,6,8-tetrayl)tetrabenzaldehyde
Precaución: for Research Use Only


Formula: C₄₄H₂₆O₄
MDL Number: MFCD31700732
CAS Number: 1415238-25-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty