Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dibromo-4-fluoroaniline 98%

Supplier: AMBEED, INC
Warning

A498497-100G A498497-25G A498497-500G A498497-5G
77292-102EA 39 USD InStock
77292-102 77292-104 77292-106 77292-108 77292-110
2,6-Dibromo-4-fluoroaniline 98%
2,6-Dibromo-4-fluoroaniline
Precaución: for Research Use Only


Formula: C₆H₄Br₂FN
MW: 268.91 g/mol
Boiling Pt: 277
Melting Pt: 68
MDL Number: MFCD00041445
CAS Number: 344-18-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty