Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(-)-1,2-Bis[(2R,5R)-2,5-dimethylphospholano]ethane(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate 98%

Supplier: AMBEED, INC
Warning

A499554-100MG A499554-1G A499554-250MG
77265-658EA 57 USD InStock
77265-658 77265-660 77265-662
(-)-1,2-Bis[(2R,5R)-2,5-dimethylphospholano]ethane(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate 98%
(-)-1,2-Bis[(2R,5R)-2,5-dimethylphospholano]ethane(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate
Precaución: for Research Use Only


Formula: C₂₃H₃₈F₃O₃P₂RhS
MDL Number: MFCD01631282
CAS Number: 177750-25-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty