Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dihydro-1-benzofuran-5-sulfonoylchloride 96%

Supplier: AMBEED, INC
Danger

A500739-1G A500739-25G A500739-5G
77253-282EA 117.42 USD InStock
77253-282 77253-284 77253-286 77253-288 77253-290
2,3-Dihydro-1-benzofuran-5-sulfonoylchloride 96%
2,3-Dihydro-1-benzofuran-5-sulfonoylchloride
Precaución: for Research Use Only


Formula: C₈H₇ClO₃S
MW: 218.66 g/mol
MDL Number: MFCD02677703
CAS Number: 115010-11-2
UN: 3261
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty