Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2R)-1-[(1R)-1-[Bis(1,1-dimethylethyl)phosphino]ethyl]-2-(di-1-naphthalenylphosphino)ferrocene ≥98%, ee 99%

Supplier: AMBEED, INC
Warning

A501052-100MG A501052-1G A501052-250MG A501052-50MG
77253-292EA 26 USD InStock
77253-292 77253-294 77253-296 77253-298
(2R)-1-[(1R)-1-[Bis(1,1-dimethylethyl)phosphino]ethyl]-2-(di-1-naphthalenylphosphino)ferrocene ≥98%, ee 99%
(2R)-1-[(1R)-1-[Bis(1,1-dimethylethyl)phosphino]ethyl]-2-(di-1-naphthalenylphosphino)ferrocene
Precaución: for Research Use Only


Formula: C₄₀H₄₄FeP₂
MDL Number: MFCD08561160
CAS Number: 849924-43-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty