Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetrakis[n-tetrachlorophthaloyl-(s)-tert-leucinato]dirhodium bis(ethyl acetate) adduct 98%

Supplier: AMBEED, INC
Warning

A501512-100MG A501512-250MG A501512-1G
77253-300EA 255 USD InStock
77253-300 77253-302 77747-027
Tetrakis[n-tetrachlorophthaloyl-(s)-tert-leucinato]dirhodium bis(ethyl acetate) adduct 98%
Tetrakis[n-tetrachlorophthaloyl-(s)-tert-leucinato]dirhodium bis(ethyl acetate) adduct
Precaución: for Research Use Only


Formula: C₅₆H₄₀Cl₁₆N₄O₁₆Rh₂
MW: 1974.17 g/mol
MDL Number: MFCD15072197
CAS Number: 1816286-21-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty