Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

MK-8998 98+%

Supplier: AMBEED, INC
Warning

A507419-10MG A507419-1MG A507419-25MG A507419-50MG A507419-5MG
77265-744EA 99 USD InStock
77265-744 77265-746 77265-748 77265-750 77265-752
MK-8998 98+%
MK-8998
Precaución: for Research Use Only


Formula: C₂₀H₂₃F₃N₂O₂
MDL Number: MFCD22572346
CAS Number: 953778-58-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty