Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,5-Dichlorophthalic acid 96%

Supplier: AMBEED, INC
Warning

A507588-10G A507588-1G A507588-25G A507588-5G
77292-156EA 30 USD InStock
77292-156 77292-158 77292-160 77292-162
4,5-Dichlorophthalic acid 96%
4,5-Dichlorophthalic acid
Precaución: for Research Use Only


Formula: C₈H₄Cl₂O₄
MDL Number: MFCD00002493
CAS Number: 56962-08-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty