Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α,α,α,α',α',α'-Hexafluoro-2,6-xylidine 95%

Supplier: AMBEED, INC
Warning

A508052-100MG A508052-1G A508052-250MG A508052-5G
77253-370EA 46 USD InStock
77253-370 77253-372 77253-374 77253-376
α,α,α,α',α',α'-Hexafluoro-2,6-xylidine 95%
α,α,α,α',α',α'-Hexafluoro-2,6-xylidine
Precaución: for Research Use Only


Formula: C₈H₅F₆N
MDL Number: MFCD01313162
CAS Number: 313-13-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty