Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Nitroso-2-naphthol 97%

Supplier: AMBEED, INC
Warning

A508318-100G A508318-1G A508318-25G A508318-500G A508318-5G
77241-566EA 122 USD InStock
77241-566 77241-568 77241-570 77241-572 77241-574
1-Nitroso-2-naphthol 97%
1-Nitroso-2-naphthol
Precaución: for Research Use Only


Formula: C₁₀H₇NO₂
MDL Number: MFCD00003884
CAS Number: 131-91-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty