Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isoguanosine 95%

Supplier: AMBEED, INC
Warning

A508462-1MG A508462-5MG
77265-770EA 15 USD InStock
77265-770 77265-772
Isoguanosine 95%
Isoguanosine
Precaución: for Research Use Only


Formula: C₁₀H₁₃N₅O₅
MDL Number: MFCD04971989
CAS Number: 1818-71-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty