Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Acetylbenzenesulfonyl chloride 97%

Supplier: AMBEED, INC
Danger

A509037-100MG A509037-10G A509037-1G A509037-250MG A509037-5G
77241-584EA 7 USD InStock
77241-584 77241-586 77241-588 77241-590 77241-592
4-Acetylbenzenesulfonyl chloride 97%
4-Acetylbenzenesulphonyl chloride
Precaución: for Research Use Only


Formula: C₈H₇ClO₃S
MDL Number: MFCD00800269
CAS Number: 1788-10-9
UN: 3261
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty