Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N-Dimethyl 3-borono-4-methylbenzenesulphonamide 98%

Supplier: AMBEED, INC
Warning

A510036-1G A510036-250MG A510036-5G
77253-398EA 207 USD InStock
77253-398 77253-400 77253-402
N,N-Dimethyl 3-borono-4-methylbenzenesulphonamide 98%
N,N-Dimethyl 3-borono-4-methylbenzenesulphonamide
Precaución: for Research Use Only


Formula: C₉H₁₄BNO₄S
MDL Number: MFCD08056363
CAS Number: 871332-99-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty