Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl 4,4'-diamino-[1,1'-biphenyl]-2,2'-dicarboxylate 97%

Supplier: AMBEED, INC
Warning

A511474-1G A511474-250MG
77241-620EA 99 USD InStock
77241-620 77241-622
Dimethyl 4,4'-diamino-[1,1'-biphenyl]-2,2'-dicarboxylate 97%
Dimethyl 4,4'-diamino-[1,1'-biphenyl]-2,2'-dicarboxylate
Precaución: for Research Use Only


Formula: C₁₆H₁₆N₂O₄
MDL Number: MFCD28359718
CAS Number: 149935-34-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty