Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Ethoxy-2-methylbenzeneboronic acid 98%

Supplier: AMBEED, INC
Warning

A511507-1G A511507-25G A511507-5G
77292-190EA 18 USD InStock
77292-190 77292-192 77292-194
4-Ethoxy-2-methylbenzeneboronic acid 98%
4-Ethoxy-2-methylbenzeneboronic acid
Precaución: for Research Use Only


Formula: C₉H₁₃BO₃
MDL Number: MFCD04115659
CAS Number: 313545-31-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty