Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 5-bromo-1,4-dimethyl-1H-pyrazole-3-carboxylate 95%

Supplier: AMBEED, INC
Warning

A511542-1G A511542-250MG
77241-624EA 682 USD InStock
77241-624 77241-626
Ethyl 5-bromo-1,4-dimethyl-1H-pyrazole-3-carboxylate 95%
Ethyl 5-bromo-1,4-dimethyl-1H-pyrazole-3-carboxylate
Precaución: for Research Use Only


Formula: C₈H₁₁BrN₂O₂
MDL Number: MFCD26381813
CAS Number: 1707370-05-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty