Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hydroxy Dynasore ≥98%

Supplier: AMBEED, INC
Danger

A511904-10MG A511904-1MG A511904-250MG A511904-50MG
77265-826EA 53 USD InStock
77265-826 77265-828 77265-830 77265-832
Hydroxy Dynasore ≥98%
Hydroxy Dynasore
Precaución: for Research Use Only


Formula: C₁₈H₁₄N₂O₅
MDL Number: MFCD23704787
CAS Number: 1256493-34-1
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty