Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-3-Carene 90% stabilised

Supplier: AMBEED, INC
Danger

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A511932-1G A511932-25G A511932-5G
77279-742EA 8 USD InStock
77279-742 77749-050 77749-051
(±)-3-Carene 90% stabilised
(±)-3-Carene
Precaución: for Research Use Only


Formula: C₁₀H₁₆
MDL Number: MFCD00001315
CAS Number: 13466-78-9
UN: 2319
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty