Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxy-3-nitrobenzonitrile 98%

Supplier: AMBEED, INC
Danger

A512188-1G A512188-250MG A512188-25G A512188-5G
77292-196EA 12 USD InStock
77292-196 77292-198 77292-200 77292-202
4-Methoxy-3-nitrobenzonitrile 98%
4-Methoxy-3-nitrobenzonitrile
Precaución: for Research Use Only


Formula: C₈H₆N₂O₃
MDL Number: MFCD00221439
CAS Number: 33224-23-6
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty