Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-O-Methyl-β-D-glucuronic acid sodium salt 98%

Supplier: AMBEED, INC
Warning

A512370-1G A512370-250MG A512370-5G
77241-632EA 14 USD InStock
77241-632 77241-634 77241-636
1-O-Methyl-β-D-glucuronic acid sodium salt 98%
1-O-Methyl-β-D-glucuronic acid sodium salt
Precaución: for Research Use Only


Formula: C₇H₁₁O₇·Na
MW: 230.148 g/mol
MDL Number: MFCD00083405
CAS Number: 134253-42-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty