Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-2-Bromoisovaleric acid 98%

Supplier: AMBEED, INC
Danger

A513543-25G A513543-5G
77292-218EA 19 USD InStock
77292-218 77292-220
(±)-2-Bromoisovaleric acid 98%
(±)-2-Bromoisovaleric acid


Formula: C₅H₉BrO₂
MDL Number: MFCD00004210
CAS Number: 565-74-2
UN: 3261
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty