Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Butyloctanoic acid 95%

Supplier: AMBEED, INC
Warning

A514029-100G A514029-10G A514029-1G A514029-25G A514029-500G
77265-858EA 67 USD InStock
77265-858 77265-860 77265-862 77265-864 77265-866
2-Butyloctanoic acid 95%
2-Butyloctanoic acid
Precaución: for Research Use Only


Formula: C₁₂H₂₄O₂
MDL Number: MFCD00661074
CAS Number: 27610-92-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty