Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(4-Chlorophenyl)-1,2,3,6-tetrahydropyridine hydrochloride 95%

Supplier: AMBEED, INC
Danger

A514169-100MG A514169-5G
77292-228EA 29 USD InStock
77292-228 77292-230
4-(4-Chlorophenyl)-1,2,3,6-tetrahydropyridine hydrochloride 95%
4-(4-Chlorophenyl)-1,2,3,6-tetrahydropyridine hydrochloride
Precaución: for Research Use Only


Formula: C₁₁H₁₃Cl₂N
MDL Number: MFCD00012753
CAS Number: 51304-61-1
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty