Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Difluoroterephthalic acid 97%

Supplier: AMBEED, INC
Warning

A515170-100MG A515170-1G A515170-250MG
77265-874EA 16 USD InStock
77265-874 77265-876 77265-878
2,5-Difluoroterephthalic acid 97%
2,5-Difluoroterephthalic acid
Precaución: for Research Use Only


Formula: C₈H₄F₂O₄
MW: 202.11 g/mol
MDL Number: MFCD00042502
CAS Number: 655-14-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty