Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dioxocyclohexanecarboxylic acid 97%

Supplier: AMBEED, INC
Warning

A516296-10G A516296-250MG A516296-25G A516296-5G A516296-1G
77279-752EA 187 USD InStock
77279-752 77279-754 77279-756 77279-758 77749-068
3,5-Dioxocyclohexanecarboxylic acid 97%
3,5-Dioxocyclohexanecarboxylic acid
Precaución: for Research Use Only


Formula: C₇H₈O₄
MDL Number: MFCD09836162
CAS Number: 42858-60-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty