Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3,5,6-Tetrachloropyridine 98%

Supplier: AMBEED, INC
Warning

A516399-10G A516399-1G A516399-25G
77253-456EA 8 USD InStock
77253-456 77253-458 77253-460
2,3,5,6-Tetrachloropyridine 98%
2,3,5,6-Tetrachloropyridine
Precaución: for Research Use Only


Formula: C₅HCl₄N
MDL Number: MFCD00092428
CAS Number: 2402-79-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty