Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-Piperazine-2-carboxylic acid dihydrochloride 98%

Supplier: AMBEED, INC
Warning

A517507-1G A517507-250MG A517507-25G
77292-254EA 6 USD InStock
77292-254 77292-256 77749-073
(R)-Piperazine-2-carboxylic acid dihydrochloride 98%
(R)-Piperazine-2-carboxylic acid dihydrochloride
Precaución: for Research Use Only


Formula: C₅H₁₂Cl₂N₂O₂
MDL Number: MFCD03840829
CAS Number: 126330-90-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty