Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-Bromophenyl)-5-phenyl-1,3,4-oxadiazole 97%

Supplier: AMBEED, INC
Warning

A520546-1G A520546-250MG
77241-680EA 35 USD InStock
77241-680 77241-682
2-(4-Bromophenyl)-5-phenyl-1,3,4-oxadiazole 97%
2-(4-Bromophenyl)-5-phenyl-1,3,4-oxadiazole
Precaución: for Research Use Only


Formula: C₁₄H₉BrN₂O
MDL Number: MFCD00050997
CAS Number: 21510-43-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty