Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dichloro-7-methylquinoline 95%

Supplier: AMBEED, INC
Danger

A521687-100MG A521687-1G A521687-250MG
77241-684EA 65 USD InStock
77241-684 77241-686 77241-688
2,4-Dichloro-7-methylquinoline 95%
2,4-Dichloro-7-methylquinoline
Precaución: for Research Use Only


Formula: C₁₀H₇Cl₂N
MDL Number: MFCD22200007
CAS Number: 102878-19-3
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty