Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2'-Dithiodianiline (thulium ionophore I) 98%

Supplier: AMBEED, INC
Danger

A523644-1G A523644-25G A523644-5G
77253-528EA 9 USD InStock
77253-528 77253-530 77253-532
2,2'-Dithiodianiline (thulium ionophore I) 98%
2,2'-Dithiodianiline (thulium ionophore I)
Precaución: for Research Use Only


Formula: C₁₂H₁₂N₂S₂
MDL Number: MFCD00007703
CAS Number: 1141-88-4
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty