Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-((5-Chloro-2-((2-methoxy-4-(4-methylpiperazin-1-yl)phenyl)amino)pyrimidin-4-yl)amino)-N-methylbenzenesulfonamide 98+%

Supplier: AMBEED, INC
Warning

A524942-10MG A524942-1MG A524942-5MG
77241-732EA 65 USD InStock
77241-732 77241-734 77241-736
2-((5-Chloro-2-((2-methoxy-4-(4-methylpiperazin-1-yl)phenyl)amino)pyrimidin-4-yl)amino)-N-methylbenzenesulfonamide 98+%
2-((5-Chloro-2-((2-methoxy-4-(4-methylpiperazin-1-yl)phenyl)amino)pyrimidin-4-yl)amino)-N-methylbenzenesulfonamide
Precaución: for Research Use Only


Formula: C₂₃H₂₈ClN₇O₃S
MDL Number: MFCD23380589
CAS Number: 761438-38-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty