Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dichlorobis(isoquinoline)-Palladium 98%

Supplier: AMBEED, INC
Warning

A526235-100MG A526235-250MG
77253-534EA 122 USD InStock
77253-534 77253-536
Dichlorobis(isoquinoline)-Palladium 98%
Dichlorobis(isoquinoline)-Palladium
Precaución: for Research Use Only


Formula: C₁₈H₁₄Cl₂N₂Pd
MDL Number: MFCD00792249
CAS Number: 23807-51-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty