Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 4-amino-2-chloronicotinate 97%

Supplier: AMBEED, INC
Danger

A528473-100MG A528473-1G A528473-250MG
77241-766EA 11 USD InStock
77241-766 77241-768 77241-770
Ethyl 4-amino-2-chloronicotinate 97%
Ethyl 4-amino-2-chloronicotinate
Precaución: for Research Use Only


Formula: C₈H₉ClN₂O₂
MDL Number: MFCD24643019
CAS Number: 1194341-67-7
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty