Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Mercapto-2-butanol (mixture of isomers) 98%

Supplier: AMBEED, INC
Danger

A528548-25G A528548-5G
77279-840EA 24 USD InStock
77279-840 77279-842
3-Mercapto-2-butanol (mixture of isomers) 98%
3-Mercapto-2-butanol (mixture of isomers)
Precaución: for Research Use Only


Formula: C₄H₁₀OS
MW: 106.18 g/mol
MDL Number: MFCD00010271
CAS Number: 54812-86-1
UN: 1993
ADR: 3 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty