Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Pyridyl trifluoromethanesulfonate 98+%

Supplier: AMBEED, INC
Danger

A529558-1G A529558-25G A529558-5G
77279-858EA 32 USD InStock
77279-858 77279-860 77279-862
2-Pyridyl trifluoromethanesulfonate 98+%
2-Pyridyl trifluoromethanesulfonate
Precaución: for Research Use Only


Formula: C₆H₄F₃NO₃S
MDL Number: MFCD00192531
CAS Number: 65007-00-3
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty