Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,5-Dibromothiophene-2-carboxaldehyde 99+%

Supplier: AMBEED, INC
Warning

A529746-1G A529746-250MG A529746-25G A529746-5G
77292-306EA 37 USD InStock
77292-306 77292-308 77292-310 77292-312
4,5-Dibromothiophene-2-carboxaldehyde 99+%
4,5-Dibromothiophene-2-carboxaldehyde
Precaución: for Research Use Only


Formula: C₅H₂Br₂OS
MW: 269.94 g/mol
MDL Number: MFCD00225242
CAS Number: 38071-22-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty