Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2-Chloroethoxy)tetrahydropyran 96%

Supplier: AMBEED, INC
Danger

A534198-1G A534198-25G A534198-5G
77241-812EA 5 USD InStock
77241-812 77241-814 77241-816
2-(2-Chloroethoxy)tetrahydropyran 96%
2-(2-Chloroethoxy)tetrahydropyran
Precaución: for Research Use Only


Formula: C₇H₁₃ClO₂
MDL Number: MFCD00142761
CAS Number: 5631-96-9
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty