Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Chlorosulfonyl)-4-fluorobenzoic acid 95%

Supplier: AMBEED, INC
Danger

A535050-10G A535050-5G A535050-25G
77279-908EA 30 USD InStock
77279-908 77279-910 77749-116
3-(Chlorosulfonyl)-4-fluorobenzoic acid 95%
3-(Chlorosulfonyl)-4-fluorobenzoic acid
Precaución: for Research Use Only


Formula: C₇H₄ClFO₄S
MW: 238.62 g/mol
MDL Number: MFCD03030336
CAS Number: 2267-40-5
UN: 3261
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty