Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

SD-169 97%

Supplier: AMBEED, INC
Warning

A538658-100MG A538658-1G A538658-250MG
77241-850EA 40 USD InStock
77241-850 77241-852 77241-854
SD-169 97%
SD-169
Precaución: for Research Use Only


Formula: C₉H₈N₂O
MW: 160.17 g/mol
MDL Number: MFCD07779486
CAS Number: 1670-87-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty