Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4'-Dichloropropiophenone 98%

Supplier: AMBEED, INC
Danger

A539741-1G A539741-25G A539741-5G
77279-922EA 9 USD InStock
77279-922 77279-924 77279-926
3,4'-Dichloropropiophenone 98%
3,4'-Dichloropropiophenone
Precaución: for Research Use Only


Formula: C₉H₈Cl₂O
MW: 203.07
Melting Pt: 51
MDL Number: MFCD00000992
CAS Number: 3946-29-0
UN: 3261
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty