Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-(+)-2,2-Dimethylcyclopropanecarboxamide 98%

Supplier: AMBEED, INC
Warning

A540472-100G A540472-10G A540472-1G A540472-25G A540472-500G A540472-5G
77292-392EA 26 USD InStock
77292-392 77292-394 77292-396 77292-398 77292-400 77292-402
(S)-(+)-2,2-Dimethylcyclopropanecarboxamide 98%
(S)-(+)-2,2-Dimethylcyclopropanecarboxamide
Precaución: for Research Use Only


Formula: C₆H₁₁NO
MDL Number: MFCD00216614
CAS Number: 75885-58-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty